Abnova
Manufacturer Code:PAB17783
Catalog # 89-119-449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human MYBPC3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of MYBPC3.
For Research Use Only
Protein Aliases: C-protein, cardiac muscle isoform; Cardiac MyBP-C; CMH4; DKFZp779E1762; FHC; MYBP-C; Myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C
Gene Aliases: CMD1MM; CMH4; FHC; LVNC10; MYBP-C; MYBPC3
UniProt ID: (Human) Q14896
Entrez Gene ID: (Human) 4607
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.