Invitrogen
Manufacturer Code:MA537716
Catalog # PIMA537716
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1702CT711.87.46 |
Immunogen | recombinant protein between 1104-1328 amino acids from human MYBBP1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AL024407 AU019902 DBP MYB binding protein 1a MYB binding protein (P160) 1a MYB binding protein 1a myb-binding protein 1A Myb-binding protein of 160 kDa Mybbp1a nuclear protein P160 P160 p160MBP p53-activated protein-2 p67MBP PAP2 PAR interacting protein PAR-interacting protein PIP; MYB binding protein (P160) 1a; Myb-binding protein 1A; p53-activated protein-2
Gene Aliases: MYBBP1A; P160; PAP2
UniProt ID: (Human) Q9BQG0
Entrez Gene ID: (Human) 10514
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.