Invitrogen
Manufacturer Code:PA596287
Catalog # PIPA596287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1104-1328 of human MYBBP1A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MYB binding protein 1a; MYB binding protein (P160) 1a; Myb-binding protein 1A; Myb-binding protein of 160 kDa; nuclear protein P160; p53-activated protein-2; PAR interacting protein; PAR-interacting protein; PIP
Gene Aliases: AL024407; AU019902; DBP; MYBBP1A; P160; p160MBP; p67MBP; PAP2
UniProt ID: (Human) Q9BQG0, (Rat) O35821, (Mouse) Q7TPV4
Entrez Gene ID: (Human) 10514, (Rat) 60571, (Mouse) 18432
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.