Novus Biologicals
Manufacturer Code:NB10061051
Catalog # NB10061051
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1278 and 1328 of human MYB binding protein (P160) 1a using the numbering given in entry NP_055335.1 (GeneID 10514). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MYB binding protein (P160) 1a; MYB binding protein (P160) 1a myb-binding protein 1A P160FLJ37886 p53-activated protein-2 PAP2; Myb-binding protein 1A; p53-activated protein-2
Gene Aliases: MYBBP1A; P160; PAP2
UniProt ID: (Human) Q9BQG0
Entrez Gene ID: (Human) 10514
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.