OriGene
Manufacturer Code:TA890056
Catalog # 50-168-4769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human MYBBP1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MYB binding protein (P160) 1a; Myb-binding protein 1A; P160 PAP2 Pol5; p53-activated protein-2
Gene Aliases: MYBBP1A; P160; PAP2
UniProt ID: (Human) Q9BQG0
Entrez Gene ID: (Human) 10514
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.