Abnova
Manufacturer Code:PAB10189
Catalog # 89-114-686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of mouse Myb. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Myb.
For Research Use Only
Protein Aliases: AI550390; c-myb; gag-myb protein; M16449; MGC18531; myb proto-oncogene protein; myeloblastosis proto-oncogene product; Proto-oncogene c-Myb; Transcriptional activator Myb; tumor-specific myb protein
Gene Aliases: AI550390; c-myb; M16449; Myb
UniProt ID: (Mouse) P06876
Entrez Gene ID: (Mouse) 17863
Molecular Function: DNA binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.