Invitrogen
Manufacturer Code:PA141578
Catalog # 01-675-197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 250-300 of human MYADM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MYADM; Myeloid up-regulated protein; myeloid upregulated protein; Myeloid-associated differentiation marker; Protein SB135
Gene Aliases: MYADM; SB135; UNQ553/PRO1110
UniProt ID: (Human) Q96S97, (Rat) Q6VBQ5
Entrez Gene ID: (Human) 91663, (Rat) 369016
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.