Invitrogen
Manufacturer Code:PA576590
Catalog # PIPA576590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human MVK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LH receptor mRNA-binding protein; Mevalonate kinase; mevalonate kinase 1; Mevalonate kinase MK KIME; MK
Gene Aliases: 2310010A05Rik; AI256848; AI414037; LRBP; MK; MVK; MVLK; POROK3
UniProt ID: (Human) Q03426, (Mouse) Q9R008
Entrez Gene ID: (Human) 4598, (Mouse) 17855
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.