Invitrogen
Manufacturer Code:PA5101144
Catalog # PIPA5101144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Mevalonate Kinase corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310010A05Rik AI256848 AI414037 KIME LH receptor mRNA-binding protein LHR mRNA binding protein Lrbp mevalonate kinase mevalonate kinase 1 MK Mvk MVLK POROK3; LH receptor mRNA-binding protein; LHR mRNA binding protein; Mevalonate kinase; mevalonate kinase 1; MK
Gene Aliases: 2310010A05Rik; AI256848; AI414037; LRBP; MK; MVK; MVLK; POROK3
UniProt ID: (Human) Q03426, (Mouse) Q9R008, (Rat) P17256
Entrez Gene ID: (Human) 4598, (Mouse) 17855, (Rat) 81727
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.