Novus Biologicals
Manufacturer Code:NBP132868
Catalog # NBP132868
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human MVK. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.36 LH receptor mRNA-binding protein LRBP mevalonate kinase mevalonate kinase (mevalonic aciduria) mevalonate kinase 1 MKFLJ96772 MVLK; LH receptor mRNA-binding protein; Mevalonate kinase; mevalonate kinase 1; MK
Gene Aliases: LRBP; MK; MVK; MVLK; POROK3
UniProt ID: (Human) Q03426
Entrez Gene ID: (Human) 4598
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.