R&D Systems
Manufacturer Code:AF8516
Catalog # AF8516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E. coli</em>-derived recombinant human MVK Met1-Lys396 Accession # Q03426 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.36 LH receptor mRNA-binding protein LRBP mevalonate kinase mevalonate kinase (mevalonic aciduria) mevalonate kinase 1 Mevalonic Aciduria MKFLJ96772 MVLK POROK3; LH receptor mRNA-binding protein; Mevalonate kinase; mevalonate kinase 1; MK
Gene Aliases: LRBP; MK; MVK; MVLK; POROK3
UniProt ID: (Human) Q03426
Entrez Gene ID: (Human) 4598
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.