Invitrogen
Manufacturer Code:PA579709
Catalog # PIPA579709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human MVD. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Diphosphomevalonate decarboxylase; Diphosphomevalonate decarboxylase Mevalonate (diphospho)decarboxylase MDDase Mevalonate pyrophosphate decarboxylase MVD MPD; MDDase; mevalonate (diphospho) decarboxylase; Mevalonate (diphospho)decarboxylase; Mevalonate pyrophosphate decarboxylase
Gene Aliases: C78718; FP17780; MDDase; MPD; MVD; Mvpd; POROK7
UniProt ID: (Human) P53602, (Rat) Q62967, (Mouse) Q99JF5
Entrez Gene ID: (Human) 4597, (Rat) 81726, (Mouse) 192156
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.