Abnova
Manufacturer Code:H00004597B02P
Catalog # 89-104-960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | MVD (NP_002452.11 a.a. &bsim 400 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human MVD protein.
For Research Use Only
Protein Aliases: Diphosphomevalonate decarboxylase; FP17780; MDDase; mevalonate (diphospho) decarboxylase; Mevalonate (diphospho)decarboxylase; Mevalonate pyrophosphate decarboxylase; MPD
Gene Aliases: FP17780; MDDase; MPD; MVD; POROK7
UniProt ID: (Human) P53602
Entrez Gene ID: (Human) 4597
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.