Invitrogen
Manufacturer Code:PA523648
Catalog # PIPA523648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 40-68 amino acids from the central region of human MUTED |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 5, muted; Biogenesis of lysosome-related organelles complex 1 subunit 5; biogenesis of organelles complex-1, subunit 5, muted; BLOC-1 subunit 5; MU protein Muted homolog; muted homolog; Protein Muted homolog
Gene Aliases: 1810074A19Rik; BLOC1S5; BLOS5; MU; MUTED
UniProt ID: (Human) Q8TDH9, (Mouse) Q3TCP2
Entrez Gene ID: (Human) 63915, (Mouse) 17828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.