Invitrogen
Manufacturer Code:PA579708
Catalog # PIPA579708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human MUT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCM; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; Methylmalonyl-CoA isomerase; Methylmalonyl-CoA mutase mitochondrial MCM; Methylmalonyl-CoA mutase, mitochondrial; methylmalonyl-Coenzyme A mutase; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase
Gene Aliases: MCM; MMUT; MUT
UniProt ID: (Human) P22033
Entrez Gene ID: (Human) 4594, (Rat) 688517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.