Invitrogen
Manufacturer Code:MA537664
Catalog # PIMA537664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1429CT456.173.44 |
Immunogen | recombinant protein from mouse Musk |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CMS9 FADS Mdk4 MGC126323 MGC126324 Mlk muscle associated receptor tyrosine kinase muscle localized kinase 2 muscle specific kinase (neural fold/somite kinase 1) muscle skeletal receptor tyrosine protein kinase Muscle skeletal receptor tyrosine-protein kinase muscle skeletal receptor tyrosine kinase muscle-specific kinase receptor muscle-specific protein kinase secretory isoform muscle-specific tyrosine kinase receptor MuSK muscle-specific tyrosine protein kinase receptor Muscle-specific tyrosine-protein kinase receptor Musk Nsk1 Nsk2 Nsk3 RP23-170A3.1; muscle localized kinase 2; muscle specific kinase (neural fold/somite kinase 1); Muscle, skeletal receptor tyrosine protein kinase; Muscle, skeletal receptor tyrosine-protein kinase; muscle-specific kinase receptor; muscle-specific protein kinase secretory isoform; muscle-specific tyrosine kinase receptor MuSK; Muscle-specific tyrosine protein kinase receptor; Muscle-specific tyrosine-protein kinase receptor; MuSK
Gene Aliases: Mdk4; Mlk; Musk; Nsk1; Nsk2; Nsk3
UniProt ID: (Mouse) Q61006, (Rat) Q62838
Entrez Gene ID: (Mouse) 18198, (Rat) 81725
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.