Proteintech
Manufacturer Code:110181AP150UL
Catalog # 50-172-6800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MUS81 Fusion Protein Ag1476 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Crossover junction endonuclease MUS81; MUS81; MUS81 endonuclease homolog; SLX3 structure-specific endonuclease subunit homolog
Gene Aliases: 1200008A18Rik; AI182501; AW045863; MUS81; SLX3
UniProt ID: (Human) Q96NY9, (Rat) Q4KM32, (Mouse) Q91ZJ0
Entrez Gene ID: (Human) 80198, (Rat) 293678, (Mouse) 71711
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.