Invitrogen
Manufacturer Code:MA535796
Catalog # PIMA535796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1518 |
Immunogen | A synthesized peptide derived from human STXBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI317162 AI326233 ANC18HA EIEE4 FLJ37475 hUNC18 MGC146482 MMS10-G Ms10g MUNC18-1 Munc-18a munc-18c NSEC1 N-Sec1 NSEC1A p67 Protein unc-18 homolog 1 Protein unc-18 homolog A rbSec1 Rb-sec1 rbSec1A rbSec1B rop RP11-56D16.3 Sec1 Stxbp1 Sxtbp1 Syntaxin-binding protein 1 UNC18 Unc18-1 UNC18A Unc-18A Unc18h; MUNC18-1; N-Sec1; neuronal SEC1; p67; Protein unc-18 homolog 1; Protein unc-18 homolog A; Syntaxin-binding protein 1; Unc-18A; Unc18-1
Gene Aliases: MUNC18-1; NSEC1; P67; RBSEC1; STXBP1; UNC18; UNC18A
UniProt ID: (Human) P61764
Entrez Gene ID: (Human) 6812
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.