Novus Biologicals
Manufacturer Code:NBP294291100UL
Catalog # NB023137
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-352 of human MUL1 (NP_078820.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C1orf166mitochondrial E3 ubiquitin ligase 1 E3 ubiquitin-protein ligase MUL1 EC 6.3.2 EC 6.3.2.- FLJ12875 GIDERP11-401M16.2 Growth inhibition and death E3 ligase MAPLE3 ubiquitin ligase mitochondrial E3 ubiquitin protein ligase 1 mitochondrial ubiquitin ligase activator of NF-kB mitochondrial ubiquitin ligase activator of NFKB 1 Mitochondrial-anchored protein ligase MULANchromosome 1 open reading frame 166 Putative NF-kappa-B-activating protein 266 RING finger protein 218 RNF218mitochondria-anchored protein ligase; E3 SUMO-protein ligase MUL1; E3 ubiquitin ligase; E3 ubiquitin-protein ligase MUL1; Growth inhibition and death E3 ligase; mitochondria-anchored protein ligase; mitochondrial E3 ubiquitin ligase 1; mitochondrial ubiquitin ligase activator of NF-kB; Mitochondrial ubiquitin ligase activator of NFKB 1; Mitochondrial-anchored protein ligase; Protein Hades; Putative NF-kappa-B-activating protein 266; RING finger protein 218; RING-type E3 ubiquitin transferase NFKB 1
Gene Aliases: C1orf166; GIDE; MAPL; MUL1; MULAN; RNF218
UniProt ID: (Human) Q969V5
Entrez Gene ID: (Human) 79594
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.