Invitrogen
Manufacturer Code:MA512178
Catalog # PIMA512178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 45M1 |
Immunogen | M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Gastric mucin; lewis B blood group antigen; Major airway glycoprotein; MUC-5AC; MUC-5AC TBM gastric mucin leB lewis B blood group antigen major airway glycoprotein mucin 5 subtypes A and C tracheobronchial/gastric mucin-5 subtype AC tracheobronchial mucin-5AC tracheobronchial mucin; mucin 5, subtypes A and C; mucin 5, subtypes A and C, tracheobronchial/gastric; mucin 5AC, oligomeric mucus/gel-forming pseudogene; Mucin-5 subtype AC, tracheobronchial; Mucin-5AC; TBM; Tracheobronchial mucin
Gene Aliases: 2210005L13Rik; leB; MGM; MUC5; MUC5AC; mucin; TBM
UniProt ID: (Human) P98088
Entrez Gene ID: (Pig) 100170143, (Human) 4586, (Mouse) 17833, (Rat) 682837
Molecular Function: cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.