Proteintech
Manufacturer Code:553431AP150UL
Catalog # 50-173-6119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ascites sialoglycoprotein; Ascites sialoglycoprotein 1; Ascites sialoglycoprotein 2; Ascites sialoglycoprotein Ascites sialoglycoprotein 1 Ascites sialoglycoprotein 2 ASGP ASGP 1 Mucin 4 beta chain ASGP 2 HSA276359 MUC 4 MUC4 Mucin 4 Testis mucin Tracheobronchial mucin; ASGP; ASGP-1; ASGP-2; MUC-4; mucin 4, tracheobronchial; Mucin-4; Mucin-4 alpha chain; Mucin-4 beta chain; Pancreatic adenocarcinoma mucin; Testis mucin; Tracheobronchial mucin
Gene Aliases: ASGP; HSA276359; MUC-4; MUC4
UniProt ID: (Human) Q99102
Entrez Gene ID: (Human) 4585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.