Invitrogen
Manufacturer Code:PA5100199
Catalog # PIPA5100199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MUC3A corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A630081J09Rik intestinal mucin Intestinal mucin-3A MUC3 MUC3A MUC-3A mucin 3 mucin 3 intestinal mucin 3A mucin 3A cell surface associated Mucin-3 mucin-3A RIKEN cDNA A630081J09 gene; intestinal mucin; Intestinal mucin-3A; MUC-3A; mucin 3, intestinal; Mucin-3A
Gene Aliases: MUC-3A; MUC3; MUC3A
UniProt ID: (Human) Q02505
Entrez Gene ID: (Mouse) 619309, (Human) 4584
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.