R&D Systems
Manufacturer Code:FAB8195S100UG
Catalog # FAB8195S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 781631 |
Immunogen | <EM>E. coli</EM>-derived recombinant human MUC4 Accession # Q99102 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: Ascites sialoglycoprotein; Ascites sialoglycoprotein 1; Ascites sialoglycoprotein 2; Ascites sialoglycoprotein ASGP HSA276359 MUC4 MUC-4 mucin 4 cell surface associated mucin 4 tracheobronchial mucin-4 Pancreatic adenocarcinoma mucin Testis mucin Tracheobronchial mucin; ASGP; ASGP-1; ASGP-2; MUC-4; mucin 4, tracheobronchial; Mucin-4; Mucin-4 alpha chain; Mucin-4 beta chain; Pancreatic adenocarcinoma mucin; Testis mucin; Tracheobronchial mucin
Gene Aliases: ASGP; HSA276359; MUC-4; MUC4
UniProt ID: (Human) Q99102
Entrez Gene ID: (Human) 4585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.