Novus Biologicals
Manufacturer Code:NBP245838
Catalog # NBP245838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E3 |
Immunogen | Full length human recombinant protein of human MUC1 (NP_001018016) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Breast carcinoma-associated antigen DF3; Breast carcinoma-associated antigen DF3 Carcinoma-associated mucin CD227 CD227 antigen DF3 antigen EMA episialin H23 antigen H23AG KL-6 MAM6 MUC-1 MUC1/ZD mucin 1 cell surface associated mucin 1 transmembrane mucin-1 Peanut-reactive urinary mucin PEMMUC-1/SEC PEMT Polymorphic epithelial mucin PUMMUC-1/X tumor associated epithelial mucin Tumor-associated epithelial membrane antigen Tumor-associated mucin; CA 15-3; Cancer antigen 15-3; Carcinoma-associated mucin; CD227; DF3 antigen; EMA; Episialin; H23 antigen; H23AG; KL-6; Krebs von den Lungen-6; Medullary cystic kidney disease, autosomal dominant; MUC-1; MUC1-beta; MUC1-CT; MUC1-NT; mucin 1, transmembrane; Mucin-1; Mucin-1 subunit alpha; Mucin-1 subunit beta; Peanut-reactive urinary mucin; PEM; PEMT; Polymorphic epithelial mucin; PUM; tumor associated epithelial mucin; Tumor-associated epithelial membrane antigen; Tumor-associated mucin
Gene Aliases: ADMCKD; ADMCKD1; CA 15-3; CD227; EMA; H23AG; KL-6; MAM6; MCD; MCKD; MCKD1; MUC-1; MUC-1/SEC; MUC-1/X; MUC1; MUC1/ZD; PEM; PEMT; PUM
UniProt ID: (Human) P15941
Entrez Gene ID: (Human) 4582
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.