Invitrogen
Manufacturer Code:PA5109995
Catalog # PIPA5109995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 207-436 of human MTUS1 (NP_065800.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI481402 Angiotensin-II type 2 receptor-interacting protein AT2 receptor-binding protein ATBP ATBP135 Atip Atip1 Atip3b ATIP4 B430010I23Rik B430305I03Rik BOS_23776 C85752 Cctsg1 Cctsg1-440 Coiled-coiled tumor suppressor gene 1 protein DKFZp586D1519 DKFZp686F20243 FLJ14295 GK1 ICIS Kiaa1288 MD44 Microtubule-associated tumor suppressor 1 Microtubule-associated tumor suppressor 1 homolog Mitochondrial tumor suppressor 1 Mitochondrial tumor suppressor 1 homolog mKIAA1288 MP44 MTSG1 MTUS1; angiotensin II AT2 receptor-interacting protein 1; Angiotensin-II type 2 receptor-interacting protein; AT2 receptor-binding protein; AT2 receptor-interacting protein; AT2 receptor-interacting protein 1; AT2R binding protein; Coiled-coiled tumor suppressor gene 1 protein; coiled-coiled tumor supressor gene 1; erythroid differentiation-related; growth factor inhibitor; Microtubule-associated tumor suppressor 1; Microtubule-associated tumor suppressor 1 homolog; Mitochondrial tumor suppressor 1; Mitochondrial tumor suppressor 1 homolog; mitochondrial tumor suppressor gene 1; transcription factor MTSG1
Gene Aliases: AI481402; ATBP; ATBP135; ATIP; Atip1; Atip3b; ATIP4; B430010I23Rik; B430305I03Rik; C85752; Cctsg1; Cctsg1-440; GK1; ICIS; KIAA1288; MD44; mKIAA1288; MP44; MTSG1; MTUS1
UniProt ID: (Human) Q9ULD2, (Mouse) Q5HZI1, (Rat) Q6IMY1
Entrez Gene ID: (Human) 57509, (Mouse) 102103, (Rat) 306487
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.