Invitrogen
Manufacturer Code:PA576049
Catalog # PIPA576049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human MTTP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABL Microsomal triglyceride transfer protein Microsomal triglyceride transfer protein large subunit MTP MTTP; microsomal triglyceride transfer protein (large polypeptide, 88kDa); microsomal triglyceride transfer protein B; Microsomal triglyceride transfer protein large subunit; microsomal triglyceride transfer protein, large subunit
Gene Aliases: 1810043K16Rik; ABL; MTP; MTTP
UniProt ID: (Human) P55157, (Mouse) O08601, (Rat) D4A1W8
Entrez Gene ID: (Human) 4547, (Mouse) 17777, (Rat) 310900
Molecular Function: apolipoprotein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.