Invitrogen
Manufacturer Code:PA563035
Catalog # PIPA563035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MTTP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABL MTP microsomal triglyceride transfer protein (large polypeptide 88kDa) microsomal triglyceride transfer protein large subunit; microsomal triglyceride transfer protein (large polypeptide, 88kDa); Microsomal triglyceride transfer protein large subunit
Gene Aliases: ABL; MTP; MTTP
UniProt ID: (Human) P55157
Entrez Gene ID: (Human) 4547
Molecular Function: apolipoprotein transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.