Invitrogen
Manufacturer Code:PA542391
Catalog # PIPA542391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MTTP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: microsomal triglyceride transfer protein (large polypeptide, 88kDa); Microsomal triglyceride transfer protein ABL MGC149819 MGC149820 MTP; microsomal triglyceride transfer protein B; Microsomal triglyceride transfer protein large subunit; microsomal triglyceride transfer protein, large subunit
Gene Aliases: 1810043K16Rik; ABL; MTP; MTTP
UniProt ID: (Human) P55157, (Mouse) O08601, (Rat) D4A1W8
Entrez Gene ID: (Rabbit) 100341961, (Human) 4547, (Dog) 487873, (Horse) 100053803, (Bovine) 280868, (Mouse) 17777, (Guinea pig) 100734244, (Rat) 310900
Molecular Function:
apolipoprotein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.