Invitrogen
Manufacturer Code:PA558251
Catalog # PIPA558251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MTRR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-methyltetrahydrofolate-homocysteine methyltransferase reductase MGC129643 MSR cblE; [methionine synthase]-cobalamin methyltransferase (cob(II)alamin reducing); AqCbl reductase; Aquacobalamin reductase; Methionine synthase reductase; methionine synthase reductase, mitochondrial; MSR
Gene Aliases: cblE; MSR; MTRR
UniProt ID: (Human) Q9UBK8
Entrez Gene ID: (Human) 4552
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.