Invitrogen
Manufacturer Code:PA544819
Catalog # PIPA544819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MTRR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-methyltetrahydrofolate-homocysteine methyltransferase reductase MGC129643 MSR cblE; [methionine synthase]-cobalamin methyltransferase (cob(II)alamin reducing); AqCbl reductase; Aquacobalamin reductase; Methionine synthase reductase; methionine synthase reductase, mitochondrial; MSR
Gene Aliases: 4732420G08; cblE; MSR; MTRR
UniProt ID: (Human) Q9UBK8, (Bovine) Q4JIJ2, (Mouse) Q8C1A3, (Rat) Q498R1
Entrez Gene ID: (Human) 4552, (Horse) 100071310, (Pig) 100516580, (Bovine) 507991, (Dog) 478623, (Rabbit) 100350605, (Guinea pig) 100732821, (Mouse) 210009, (Rat) 290947
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.