OriGene
Manufacturer Code:CF501005
Catalog # 50-166-0044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E3 |
Immunogen | Full length human recombit protein of human MTRF1L produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HMRF1L MRF1L mtRF1a; mitochondrial release factor 1 like; Mitochondrial translational release factor 1-like; mtRF1a; Peptide chain release factor 1-like, mitochondrial
Gene Aliases: HMRF1L; MRF1L; MTRF1A; MTRF1L
UniProt ID: (Human) Q9UGC7
Entrez Gene ID: (Human) 54516
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.