Proteintech
Manufacturer Code:166941AP150UL
Catalog # 50-173-1120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTRF1L Fusion Protein Ag10081 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mitochondrial release factor 1 like; Mitochondrial translational release factor 1-like; MRF1L mtRF1a MTRF1L; mtRF1a; Peptide chain release factor 1-like, mitochondrial
Gene Aliases: 9130004K12Rik; HMRF1L; MRF1L; MTRF1A; MTRF1L
UniProt ID: (Human) Q9UGC7, (Mouse) Q8BJU9, (Rat) Q4V7E5
Entrez Gene ID: (Human) 54516, (Mouse) 108853, (Rat) 361473
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.