Proteintech
Manufacturer Code:115811AP150UL
Catalog # 50-172-7284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTRF1 Fusion Protein Ag2170 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mitochontrial peptide chain release factor 1; MRF 1 MRF1 MtRF 1 MTRF1 MTTRF1 RF1; MRF-1; mtRF-1; Peptide chain release factor 1, mitochondrial
Gene Aliases: A830062K05Rik; MRF1; MtRF-1; MTRF1; MTTRF1; RF1
UniProt ID: (Human) O75570, (Mouse) Q8K126
Entrez Gene ID: (Human) 9617, (Mouse) 211253
Molecular Function:
RNA binding protein
nucleic acid binding
translation factor
translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.