Invitrogen
Manufacturer Code:PA5115599
Catalog # PIPA5115599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MTR corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-methyltetrahydrofolate--homocysteine methyltransferase; 5-methyltetrahydrofolate-homocysteine methyltransferase 1; 5-methyltetrahydrofolate-homocysteine methyltransferase 5-methyltetrahydrofolate--homocysteine methyltransferase 5-methyltetrahydrofolate-homocysteine methyltransferase 1 AI894170 cblG cobalamin-dependent methionine synthase D830038K18Rik FLJ33168 FLJ43216 FLJ45386 HMAG Methionine synthase methioninesynthase MS MTR Vitamin-B12 dependent methionine synthase; Cobalamin-dependent methionine synthase; Methionine synthase; MS; Vitamin-B12 dependent methionine synthase
Gene Aliases: AI894170; cblG; D830038K18Rik; HMAG; methioninesynthase; MS; MTR
UniProt ID: (Human) Q99707, (Rat) Q9Z2Q4, (Mouse) A6H5Y3
Entrez Gene ID: (Human) 4548, (Rat) 81522, (Mouse) 238505
Molecular Function:
decarboxylase
lyase
methyltransferase
oxidoreductase
reductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.