Abnova
Manufacturer Code:H00136319B01P
Catalog # 89-002-867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTPN (AAH28093 1 a.a. ~ 118 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human MTPN protein.
For Research Use Only
Protein Aliases: FLJ31098; FLJ99857; GCDP; granule cell differentiation protein; Myotrophin; Protein V-1; V-1
Gene Aliases: GCDP; MTPN; V-1
UniProt ID: (Human) P58546
Entrez Gene ID: (Human) 136319
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.