OriGene
Manufacturer Code:CF805930
Catalog # 50-166-7620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1766-2144 of human MTOR produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FRAP FRAP1 FRAP2 RAFT1 RAPT1 SKS; Mammalian target of rapamycin; Mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mTOR; Rapamycin and FKBP12 target 1; rapamycin associated protein FRAP2; Rapamycin target protein 1; Serine/threonine-protein kinase mTOR
Gene Aliases: FRAP; FRAP1; FRAP2; MTOR; RAFT1; RAPT1; SKS
UniProt ID: (Human) P42345
Entrez Gene ID: (Human) 2475
Molecular Function: kinase non-receptor serine/threonine protein kinase nucleic acid binding nucleotide kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.