Invitrogen
Manufacturer Code:PA544221
Catalog # PIPA544221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MTO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homolog of yeast Mto1; mitochondrial MTO1-3; mitochondrial translation optimization 1 homolog; Mitochondrial translation optimization 1 homolog (S. cerevisiae) CGI-02; Protein MTO1 homolog, mitochondrial
Gene Aliases: 2310039H01Rik; 5730419A02Rik; CGI-02; COXPD10; MTO1
UniProt ID: (Human) Q9Y2Z2, (Mouse) Q923Z3
Entrez Gene ID: (Horse) 100065059, (Rabbit) 100328659, (Human) 25821, (Bovine) 535768, (Pig) 100515594, (Dog) 474967, (Mouse) 68291, (Rat) 300852, (Guinea pig) 100714255
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.