Invitrogen
Manufacturer Code:PA5100072
Catalog # PIPA5100072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MTO1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310039H01Rik 5730419A02Rik CGI02 CGI-02 COXPD10 homolog of yeast Mto1 mitochondrial MTO1-3 mitochondrial translation optimization 1 homolog mitochondrial translation optimization 1 homolog (S. cerevisiae) mitochondrial tRNA translation optimization 1 MTO1 Protein MTO1 homolog mitochondrial; homolog of yeast Mto1; mitochondrial MTO1-3; mitochondrial translation optimization 1 homolog; Protein MTO1 homolog, mitochondrial
Gene Aliases: 2310039H01Rik; 5730419A02Rik; CGI-02; COXPD10; MTO1
UniProt ID: (Human) Q9Y2Z2, (Mouse) Q923Z3
Entrez Gene ID: (Human) 25821, (Mouse) 68291, (Rat) 300852
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.