Invitrogen
Manufacturer Code:PA5102107
Catalog # PIPA5102107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MTR1B corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGQTL2 Mel1b mel1b melatonin receptor Mel-1b melatonin receptor mel1b receptor mel-1B-R Mel1b-r melatonin receptor 1B melatonin receptor 1B variant b melatonin receptor MEL1B melatonin receptor type 1B Mt2 MT2 melatonin receptor MTNR1B; Mel-1B-R; mel1b melatonin receptor; mel1b receptor; melatonin receptor 1B variant b; melatonin receptor MEL1B; Melatonin receptor type 1B
Gene Aliases: FGQTL2; MEL-1B-R; Mel1b-r; MT2; MTNR1B
UniProt ID: (Human) P49286, (Rat) P49287
Entrez Gene ID: (Human) 4544, (Rat) 192646
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.