Abnova
Manufacturer Code:PAB16494
Catalog # 89-118-366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human MTNR1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of MTNR1B.
For Research Use Only
Protein Aliases: MEL-1B-R; mel1b receptor; melatonin receptor 1B variant b; melatonin receptor MEL1B; Melatonin receptor type 1B; MT2
Gene Aliases: FGQTL2; MEL-1B-R; MT2; MTNR1B
UniProt ID: (Human) P49286
Entrez Gene ID: (Human) 4544
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.