Proteintech
Manufacturer Code:134881AP150UL
Catalog # 50-172-8779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTMR4 Fusion Protein Ag4310 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FYVE domain-containing dual specificity protein phosphatase 2; FYVE DSP2 KIAA0647 MTMR4 myotubularin related protein 4 ZFYVE11; FYVE zinc finger phosphatase; FYVE-DSP2; Myotubularin-related protein 4; Zinc finger FYVE domain-containing protein 11; zinc finger, FYVE domain containing 11
Gene Aliases: AA596759; ESTM44; FYVE-DSP2; KIAA0647; mKIAA0647; MTMR4; ZFYVE11
UniProt ID: (Human) Q9NYA4, (Mouse) Q91XS1
Entrez Gene ID: (Human) 9110, (Mouse) 170749
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.