Novus Biologicals
Manufacturer Code:NBP246035
Catalog # NBP246035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1F10 |
Immunogen | Full length human recombinant protein of human MTMR2(NP_057240) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CMT4B CMT4B1 EC 3.1.3 EC 3.1.3.2 EC 3.1.3.48 KIAA1073EC 3.1.3.- myotubularin related protein 2 myotubularin-related protein 2; Myotubularin-related protein 2; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; phosphatidylinositol-3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase; phosphoinositide-3-phosphatase
Gene Aliases: CMT4B; CMT4B1; KIAA1073; MTMR2
UniProt ID: (Human) Q13614
Entrez Gene ID: (Human) 8898
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.