Proteintech
Manufacturer Code:149731AP150UL
Catalog # 50-172-9877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTMR14 Fusion Protein Ag6883 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C3orf29 hEDTP HJUMPY MTMR14 NS5ATP4ABP1; egg-derived tyrosine phosphatase homolog; HCV NS5A-transactivated protein 4 splice variant A-binding protein 1; hJumpy; jumpy; mJumpy; Myotubularin-related protein 14; NS5ATP4ABP1
Gene Aliases: 1110061O04Rik; AW553738; C3orf29; C76151; MTMR14; RGD1304842
UniProt ID: (Human) Q8NCE2, (Mouse) Q8VEL2
Entrez Gene ID: (Human) 64419, (Mouse) 97287, (Rat) 312634
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.