Invitrogen
Manufacturer Code:PA531375
Catalog # PIPA531375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 453 and 656 of Human MTMR12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-PAP; 3-PAP PIP3AP 3-phosphatase adapter protein 3-phosphatase adapter subunit myotubularin-related protein 12 phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit phosphatidylinositol-3-phosphate asso; 3-phosphatase adapter protein; 3-phosphatase adapter subunit; Inactive phosphatidylinositol 3-phosphatase 12; Myotubularin-related protein 12; Phosphatidylinositol 3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol-3 phosphate 3-phosphatase adapter subunit; phosphatidylinositol-3 phosphate 3-phosphatase adaptor subunit; phosphatidylinositol-3-phosphatase associated protein; phosphatidylinositol-3-phosphate associated protein
Gene Aliases: 3-PAP; 3Pap; 4932703C11; C730015A02Rik; im:7160376; KIAA1682; mKIAA1682; MTMR12; PIP3AP; si:dkey-207e22.3
UniProt ID: (Human) Q9C0I1, (Mouse) Q80TA6, (Zebrafish) A2BGG1
Entrez Gene ID: (Human) 54545, (Mouse) 268783, (Zebrafish) 569130
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.