Invitrogen
Manufacturer Code:PA517972
Catalog # PIPA517972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SSPSQMMPHVQTHF) corresponding to the C-terminus amino acids of MTM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MTM1 myotubular myopathy 1 CNM MTMX XLMTM myotubularin myotubularin 1; Myotubularin; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase
Gene Aliases: AF073996; CG2; CNM; mKIAA4176; Mtm; MTM1; MTMX; XLMTM
UniProt ID: (Human) Q13496, (Mouse) Q9Z2C5
Entrez Gene ID: (Human) 4534, (Mouse) 17772
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.