Proteintech
Manufacturer Code:139241AP150UL
Catalog # 50-172-9095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTM1 Fusion Protein Ag4940 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG2 CNM MTM1 MTMX Myotubularin myotubularin 1 XLMTM; Myotubularin; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase; Phosphatidylinositol-3-phosphate phosphatase; X-linked myotubular myopathy gene 1
Gene Aliases: AF073996; CG2; CNM; mKIAA4176; Mtm; MTM1; MTMX; XLMTM
UniProt ID: (Human) Q13496, (Rat) Q6AXQ4, (Mouse) Q9Z2C5
Entrez Gene ID: (Human) 4534, (Rat) 288762, (Mouse) 17772
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.