Proteintech
Manufacturer Code:131141AP150UL
Catalog # 50-172-8499
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MTHFS Fusion Protein Ag3781 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5, 10-methenyltetrahydrofolate synthetase; 5,10-methenyl-tetrahydrofolate synthetase; 5-formyltetrahydrofolate cyclo-ligase; HsT19268 Methenyl THF synthetase MTHFS; methenyl-THF synthetase; MTHFS
Gene Aliases: 1110034I12Rik; 2310020H23Rik; AI119695; HsT19268; MTHFS
UniProt ID: (Human) P49914, (Mouse) Q9D110
Entrez Gene ID: (Human) 10588, (Rat) 300886, (Mouse) 107885
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.