Invitrogen
Manufacturer Code:PA544491
Catalog # PIPA544491
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MTHFD2L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Methenyltetrahydrofolate cyclohydrolase; methylenetetrahydrofolate dehydrogenase 2-like; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; Probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like; putative bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like protein; tetrahydrofolate dehydrogenase/cyclohydrolase
Gene Aliases: 1110019K23Rik; C630010D07Rik; MTHFD2L; RGD1310879; wu:fi14c12; zgc:63479
UniProt ID: (Human) Q9H903, (Rat) D3ZUA0, (Mouse) D3YZG8
Entrez Gene ID: (Dog) 482197, (Bovine) 536269, (Pig) 100525706, (Horse) 100056425, (Human) 441024, (Rabbit) 100344170, (Rat) 305248, (Mouse) 665563, (Zebrafish) 327496
Molecular Function:
dehydrogenase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.