Invitrogen
Manufacturer Code:PA5113383
Catalog # PIPA5113383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2 protein (136-268AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110019K23Rik BOS_6819 C630010D07Rik FLJ13105 LOC100344170 Methenyltetrahydrofolate cyclohydrolase methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 like methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like methylenetetrahydrofolate dehydrogenase 2-like MGC45532 MGC72244 Mthfd2l MTHFD2-like NAD-dependent methylenetetrahydrofolate dehydrogenase NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein Probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2 probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2-like RGD1310879 tetrahydrofolate dehydrogenase/cyclohydrolase Unknown (protein for MGC:63479) wu:fi14c12 zgc:63479; Methenyltetrahydrofolate cyclohydrolase; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; Probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; tetrahydrofolate dehydrogenase/cyclohydrolase
Gene Aliases: 1110019K23Rik; C630010D07Rik; MTHFD2L
UniProt ID: (Human) Q9H903, (Mouse) D3YZG8
Entrez Gene ID: (Human) 441024, (Mouse) 665563
Molecular Function:
dehydrogenase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.