Abnova
Manufacturer Code:PAB7581
Catalog # 89-133-386
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human MTHFD2L. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of MTHFD2L.
For Research Use Only
Protein Aliases: FLJ13105; Methenyltetrahydrofolate cyclohydrolase; MGC45532; MGC72244; MTHFD2-like; NAD-dependent methylenetetrahydrofolate dehydrogenase; NADP-dependent methylenetetrahydrofolate dehydrogenase 2-like protein; Probable bifunctional methylenetetrahydrofolate dehydrogenase/cyclohydrolase 2; tetrahydrofolate dehydrogenase/cyclohydrolase
Gene Aliases: MTHFD2L
UniProt ID: (Human) Q9H903
Entrez Gene ID: (Human) 441024
Molecular Function: dehydrogenase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.